Assay Details
| Gene Symbol: Rbm17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152824.1 | NP_690037.1 | 4 - 5 | 532 | 90 | |
| GenBank mRNA | AF083384.1 | - | 3 - 4 | 402 | 90 | |
| AK012296.1 | - | 4 - 5 | 477 | 90 | ||
| BC034896.1 | - | 4 - 5 | 532 | 1446602 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbm17
Genomic Map