Assay Details
| Gene Symbol: Spib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019866.1 | NP_063919.1 | 4 - 5 | 374 | 139 | |
| GenBank mRNA | BC141118.1 | - | 4 - 5 | 374 | 9055730 | 139 |
More Information
Updated Mapping Information
Set Membership: |
|

Spib
Genomic Map