Assay Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019985.3 | NP_064369.1 | 1 - 2 | 384 | 88 | |
| GenBank mRNA | AF201457.1 | - | 1 - 2 | 95 | 88 | |
| BC064054.1 | - | 1 - 2 | 384 | 574821 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clec1b
Genomic Map