Assay Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204239.1 | NP_001191168.1 | 2 - 3 | 511 | 121 | |
| NM_019985.3 | NP_064369.1 | 3 - 4 | 607 | 121 | ||
| GenBank mRNA | AF201457.1 | - | 3 - 4 | 318 | 121 | |
| AK007925.1 | - | 3 - 4 | 587 | 121 | ||
| BC064054.1 | - | 3 - 4 | 607 | 574821 | 121 | |
| EF070191.1 | - | 2 - 3 | 201 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clec1b
Genomic Map