Assay Details
| Gene Symbol: Pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026784.3 | NP_081060.2 | 3 - 4 | 568 | 90 | |
| NM_027348.1 | NP_081624.1 | 3 - 4 | 537 | 90 | ||
| GenBank mRNA | AK003607.1 | - | 3 - 4 | 371 | 90 | |
| AK013477.1 | - | 3 - 4 | 537 | 90 | ||
| AK150483.1 | - | 3 - 4 | 487 | 90 | ||
| AK168358.1 | - | 3 - 4 | 463 | 90 | ||
| BC028659.1 | - | 2 - 3 | 356 | 1247653 | 90 | |
More Information
Set Membership: |
|

Pmvk
Genomic Map