Assay Details
| Gene Symbol: Olfr1225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146891.2 | NP_667102.2 | 1 | 924 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126532 | NM_146891.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126532 | NM_146891.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380824 | BC145849.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380824 | NM_146891.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380824 | BC145849.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380824 | NM_146891.2 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01513394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01198804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02154701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01356097 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484800 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484799 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200034020.mm.1 | NM_146891.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200034020.mm.1 | NM_146891.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200034020.mm.1 | TC0200004046.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 471643 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 103130408 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3800161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240818 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232321 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240818 | - | - |
| mouse-6 v1.1 (Illumina) | scl4288.1.1_271-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_22129100-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800161 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103130408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4288.1.1_271-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103130408 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240818 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240818 | - | - |

Olfr1225
Genomic Map