Assay Details
| Gene Symbol: Zkscan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130529.2 | NP_001124001.1 | 5 - 6 | 1281 | 98 | |
| NM_001291014.1 | NP_001277943.1 | 5 - 6 | 1035 | 98 | ||
| NM_172941.4 | NP_766529.3 | 5 - 6 | 1328 | 98 | ||
| GenBank mRNA | AK051711.1 | - | 5 - 6 | 617 | 98 | |
| AK139346.1 | - | 5 - 6 | 1571 | 98 | ||
| AK155196.1 | - | 5 - 6 | 858 | 98 | ||
| AK155490.1 | - | 5 - 6 | 910 | 98 | ||
| AK158448.1 | - | 5 - 6 | 866 | 98 | ||
| AK170924.1 | - | 5 - 6 | 858 | 98 | ||
| AK170989.1 | - | 5 - 6 | 863 | 98 | ||
| AK171190.1 | - | 5 - 6 | 910 | 98 | ||
| AY302242.1 | - | 4 - 5 | 809 | 98 | ||
| BC040205.1 | - | 4 - 5 | 813 | 4947290 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zkscan17
Genomic Map