Assay Details
Gene Symbol: Zkscan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130529.2 | NP_001124001.1 | 8 - 9 | 1598 | 94 | |
NM_001291014.1 | NP_001277943.1 | 8 - 9 | 1352 | 94 | ||
NM_172941.4 | NP_766529.3 | 8 - 9 | 1645 | 94 | ||
GenBank mRNA | AK051711.1 | - | 8 - 9 | 934 | 94 | |
AK155196.1 | - | 8 - 9 | 1175 | 94 | ||
AK155490.1 | - | 8 - 9 | 1227 | 94 | ||
AK170924.1 | - | 8 - 9 | 1175 | 94 | ||
AK170989.1 | - | 8 - 9 | 1180 | 94 | ||
AK171190.1 | - | 8 - 9 | 1227 | 94 | ||
AK177146.1 | - | 1 - 2 | 58 | 94 | ||
AK210528.1 | - | 1 - 2 | 91 | 94 | ||
AY302242.1 | - | 7 - 8 | 1126 | 94 | ||
BC040205.1 | - | 7 - 8 | 1130 | 4947290 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
