Assay Details
| Gene Symbol: Pramel7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178250.2 | NP_839981.1 | 2 - 3 | 862 | 117 | |
| GenBank mRNA | AF490343.1 | - | 2 - 3 | 862 | 117 | |
| BC052825.1 | - | 2 - 3 | 873 | 30030731 | 117 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078660 | AF490343.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078660 | BC052825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078660 | NM_178250.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 101815 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131856 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078660 | AF490343.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078660 | BC052825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078660 | NM_178250.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21406158 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131856 | - | - |
| Whole Mouse Genome G4122A | A_52_P200451 | AF490343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P200451 | BC052825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P200451 | NM_178250.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116862 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01295287 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01727248 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01295289 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01295284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21406123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01981827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01289058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01727248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01295284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01184646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01489894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01931759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02096829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01230991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01184638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01295287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02034958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02034961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01184637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01295289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01489898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02254407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01184635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21406101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484672 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484671 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439809_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439810_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484669 | - | - |
| Mouse Genome 430 2.0 Array | 1439810_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1439810_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1439809_at | - | - |
| Mouse Genome 430 2.0 Array | 1439809_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 347712_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 347712_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200033859.mm.1 | NM_178250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033859.mm.1 | NM_178250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033859.mm.1 | TC0200003980.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200033860.mm.1 | NM_178250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033860.mm.1 | NM_178250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033860.mm.1 | TC0200003980.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200033858.mm.1 | NM_178250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033858.mm.1 | NM_178250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033858.mm.1 | TC0200003980.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439810_s_at | - | - |
| Mouse Expression Set 430 | 1439810_PM_s_at | - | - |
| Mouse Expression Set 430 | 1439809_PM_at | - | - |
| Mouse Expression Set 430 | 1439809_at | - | - |
| Murine Genome U74v2 Set | 93245_PM_at | - | - |
| Murine Genome U74v2 Set | 93245_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439809_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439809_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439810_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439810_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439810_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439809_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439809_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439810_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 342598 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6860347 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860347 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256946 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256946 | - | - |
| mouse-6 v1.1 (Illumina) | scl19043.4.1_203-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860347 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19043.4.1_203-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860347 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256946 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256946 | - | - |

Pramel7
Genomic Map