Assay Details
| Gene Symbol: Akr1b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008012.1 | NP_032038.1 | 4 - 5 | 492 | 123 | |
| GenBank mRNA | AK207771.1 | - | 1 - 2 | 61 | 123 | |
| BC005789.1 | - | 4 - 5 | 481 | 3603424 | 123 | |
| U04204.1 | - | 4 - 5 | 492 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akr1b8
Genomic Map