Assay Details
| Gene Symbol: Akr1b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008012.1 | NP_032038.1 | 9 - 10 | 969 | 127 | |
| GenBank mRNA | BC005789.1 | - | 9 - 10 | 958 | 3603424 | 127 |
| U04204.1 | - | 9 - 10 | 969 | 127 | ||
More Information
Set Membership: |
|

Akr1b8
Genomic Map