Assay Details
| Gene Symbol: Drg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021354.1 | NP_067329.1 | 7 - 8 | 689 | 75 | |
| GenBank mRNA | AJ243590.1 | - | 7 - 8 | 689 | 75 | |
| AK012988.1 | - | 7 - 8 | 657 | 75 | ||
| BC082564.1 | - | 7 - 8 | 645 | 30551418 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Drg2
Genomic Map