| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700036132.mm.1 |
NM_030240.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036132.mm.1 |
NM_030240.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036132.mm.1 |
TC0700004661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700039197.mm.1 |
NM_030240.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700039197.mm.1 |
NM_030240.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700039197.mm.1 |
TC0700004661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036129.mm.1 |
NM_030240.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036129.mm.1 |
NM_030240.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036129.mm.1 |
TC0700004661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036133.mm.1 |
NM_030240.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036133.mm.1 |
NM_030240.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036133.mm.1 |
TC0700004661.mm.1 |
MTA TC |