Assay Details
| Gene Symbol: Aldh3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112725.1 | NP_001106196.1 | 5 - 6 | 740 | 89 | |
| NM_007436.2 | NP_031462.2 | 5 - 6 | 872 | 89 | ||
| GenBank mRNA | U12785.1 | - | 5 - 6 | 869 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh3a1
Genomic Map