Assay Details
| Gene Symbol: Aldh3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112725.1 | NP_001106196.1 | 6 - 7 | 849 | 86 | |
| NM_007436.2 | NP_031462.2 | 6 - 7 | 981 | 86 | ||
| GenBank mRNA | AF072815.1 | - | 6 - 7 | 887 | 86 | |
| AK144241.1 | - | 6 - 7 | 852 | 86 | ||
| BC144946.1 | - | 6 - 7 | 881 | 9053494 | 86 | |
| BC150677.1 | - | 6 - 7 | 881 | 9087588 | 86 | |
| EL959139.1 | - | 1 - 2 | 128 | 86 | ||
| EL963960.1 | - | 2 - 3 | 329 | 86 | ||
| U12785.1 | - | 6 - 7 | 978 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh3a1
Genomic Map