Assay Details
| Gene Symbol: Rasa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009025.2 | NP_033051.2 | 9 - 10 | 926 | 128 | |
| GenBank mRNA | AK053353.1 | - | 9 - 10 | 922 | 128 | |
| AK141511.1 | - | 9 - 10 | 933 | 128 | ||
| AK147896.1 | - | 9 - 10 | 894 | 128 | ||
| AK177970.1 | - | 2 - 3 | 127 | 128 | ||
| BC057300.1 | - | 9 - 10 | 917 | 6821254 | 128 | |
| BC060195.1 | - | 9 - 10 | 941 | 128 | ||
| BC068297.1 | - | 9 - 10 | 790 | 30102545 | 128 | |
| U20238.1 | - | 9 - 10 | 911 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rasa3
Genomic Map