Assay Details
| Gene Symbol: Usp17la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007887.2 | NP_031913.1 | 3 | 1241 | 145 | |
| GenBank mRNA | AY148453.1 | - | 2 | 1070 | 145 | |
| BC140988.1 | - | 3 | 1270 | 9055600 | 145 | |
| U41636.1 | - | 3 | 1253 | 145 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
ubiquitin-dependent protein catabolic process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity
proteolysis
ubiquitin-dependent protein catabolic process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002795 | U41636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002795 | NM_007887.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002792 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002792 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002795 | U41636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002795 | NM_007887.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302273 | U41636.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05983681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05983681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06433592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06289501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389634 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555897 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420773_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566387 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555894 | - | - |
| Mouse Genome 430 2.0 Array | 1420773_at | - | - |
| Mouse Genome 430 2.0 Array | 1420773_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700012916.mm.1 | TC0700001535.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012916.mm.1 | NM_007887.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012916.mm.1 | NM_007887.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012919.mm.1 | TC0700001535.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012919.mm.1 | NM_007887.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012919.mm.1 | NM_007887.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012914.mm.1 | TC0700001535.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012918.mm.1 | TC0700001535.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012918.mm.1 | NM_007887.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012918.mm.1 | NM_007887.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012920.mm.1 | TC0700001535.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012920.mm.1 | NM_007887.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012920.mm.1 | NM_007887.2 | Refseq |
| Mouse Expression Set 430 | 1420773_at | - | - |
| Mouse Expression Set 430 | 1420773_PM_at | - | - |
| Murine Genome U74v2 Set | 100680_at | - | - |
| Murine Genome U74v2 Set | 100680_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420773_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420773_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420773_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420773_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 534356 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4060403 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6520215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6520215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222276 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222276 | - | - |
| mouse-6 v1.1 (Illumina) | scl0013531.2_165-S | - | - |
| mouse-6 v1.1 (Illumina) | scl013531.1_184-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520215 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013531.1_184-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013531.2_165-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520215 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1222276 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222276 | - | - |

Usp17la
Genomic Map