Assay Details
| Gene Symbol: Klk1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010645.2 | NP_034775.1 | 3 | 340 | 100 | |
| GenBank mRNA | AK089370.1 | - | 3 | 358 | 100 | |
| BC026378.1 | - | 3 | 340 | 4165617 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
tissue kallikrein-kinin cascade
bradykinin biosynthetic process
regulation of systemic arterial blood pressure
left ventricular cardiac muscle tissue morphogenesis
proteolysis
vasodilation
cardiac muscle contraction
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity
tissue kallikrein-kinin cascade
bradykinin biosynthetic process
regulation of systemic arterial blood pressure
left ventricular cardiac muscle tissue morphogenesis
proteolysis
vasodilation
cardiac muscle contraction
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200494 | NM_010645.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200494 | AK089370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200494 | BC026378.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200494 | NM_010645.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200494 | AK089370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200494 | BC026378.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603377 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55867 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200494 | NM_010645.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200494 | AK089370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200494 | BC026378.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200494 | NM_010645.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200494 | AK089370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200494 | BC026378.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200494 | NM_010645.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200494 | AK089370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200494 | BC026378.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200494 | NM_010645.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200494 | AK089370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200494 | BC026378.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25084971 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178782 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178774 | - | - |
| Whole Mouse Genome G4122A | A_51_P200494 | NM_010645.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200494 | AK089370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200494 | BC026378.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200494 | NM_010645.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200494 | AK089370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200494 | BC026378.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06178774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06223296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25084959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06074379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06621231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06019573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06474249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06074369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06103406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06074370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25084949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06659878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06223302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06621224 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552565 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552564 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420701_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552562 | - | - |
| Mouse Genome 430 2.0 Array | 1420701_at | - | - |
| Mouse Genome 430 2.0 Array | 1420701_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16623_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16623_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700006362.mm.1 | TC0700000784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006362.mm.1 | NM_010645.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006358.mm.1 | TC0700000784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006358.mm.1 | NM_010645.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006360.mm.1 | TC0700000784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006360.mm.1 | NM_010645.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006359.mm.1 | TC0700000784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006359.mm.1 | NM_010645.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006361.mm.1 | TC0700000784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006361.mm.1 | NM_010645.3 | Refseq |
| Mouse Expression Set 430 | 1420701_PM_at | - | - |
| Mouse Expression Set 430 | 1420701_at | - | - |
| Murine Genome U74v2 Set | 94716_f_at | - | - |
| Murine Genome U74v2 Set | 95775_f_at | - | - |
| Murine Genome U74v2 Set | 100681_PM_f_at | - | - |
| Murine Genome U74v2 Set | 100681_f_at | - | - |
| Murine Genome U74v2 Set | 95775_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94716_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420701_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420701_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420701_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420701_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 885928 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6040369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102900519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2661092 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247708 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2661092 | - | - |
| mouse-6 v1.1 (Illumina) | scl016623.5_126-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_31981660-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102900519 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016623.5_126-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040369 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102900519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256119 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2989749 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256119 | - | - |

Klk1b1
Genomic Map