Assay Details
| Gene Symbol: Zfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177565.3 | NP_808233.1 | 2 | 584 | 76 | |
| GenBank mRNA | AK045753.1 | - | 2 | 585 | 76 | |
| BC096463.1 | - | 2 | 566 | 6466366 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902561 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902559 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902562 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613643 | AK045753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613643 | BC032259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613643 | BC096463.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613643 | NM_177565.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613643 | AK045753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613643 | BC032259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613643 | BC096463.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P613643 | NM_177565.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364763 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23450 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173205 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613643 | AK045753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613643 | BC032259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613643 | BC096463.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613643 | NM_177565.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613643 | AK045753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613643 | BC032259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613643 | BC096463.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P613643 | NM_177565.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310715 | AK045753.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310715 | BC096463.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310715 | NM_177565.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27902571 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173205 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07389524 | - | - |
| Whole Mouse Genome G4122A | A_52_P613643 | AK045753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P613643 | BC032259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P613643 | BC096463.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P613643 | NM_177565.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P613643 | AK045753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P613643 | BC032259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P613643 | BC096463.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P613643 | NM_177565.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310715 | AK045753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310715 | BC096463.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310715 | NM_177565.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132607 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173205 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09664706 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09664699 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09664709 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09664696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09664702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902559 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27902561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08993673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09301154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09657357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09649072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09625391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09468944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09657359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09625397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09566745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09566741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09625394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09649075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09052817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09337716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09195295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09657361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27902582 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10377986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377985 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439563_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460076_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10377984 | - | - |
| Mouse Genome 430 2.0 Array | 1439563_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460076_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447338_at | - | - |
| Mouse Genome 430 2.0 Array | 1447338_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460076_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439563_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 193043_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 193043_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100008672.mm.1 | NM_177565.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008672.mm.1 | NM_177565.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008672.mm.1 | TC1100000966.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100008670.mm.1 | NM_177565.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008670.mm.1 | NM_177565.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008670.mm.1 | TC1100000966.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439563_at | - | - |
| Mouse Expression Set 430 | 1439563_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439563_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439563_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 857251 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1340138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340138 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221886 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221886 | - | - |
| mouse-6 v1.1 (Illumina) | scl41325.2_569-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340138 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41325.2_569-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340138 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221886 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221886 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41325.2_569-S | - | - |

Zfp3
Genomic Map