Assay Details
| Gene Symbol: Ifitm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112715.1 | NP_001106186.1 | 2 | 615 | 107 | |
| NM_026820.3 | NP_081096.3 | 3 | 589 | 107 | ||
| GenBank mRNA | AK004121.1 | - | 2 | 478 | 107 | |
| AK169960.1 | - | 2 | 460 | 107 | ||
| AV136334.1 | - | 1 | 107 | |||
| BC027285.1 | - | 3 | 805 | 3492212 | 107 | |
| BC090258.1 | - | 2 | 443 | 4982747 | 107 | |
| BC090972.1 | - | 2 | 543 | 30297959 | 107 | |
| CR519995.1 | - | 1 | 107 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146254 | BC090972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146254 | NM_001112715.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146254 | NM_026820.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634279 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158557 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087265 | BC090258.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146254 | BC090972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146254 | NM_001112715.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146254 | NM_026820.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585964 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158557 | - | - |
| Whole Mouse Genome G4122A | A_52_P541802 | BC027285.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541802 | BC090258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541802 | BC090972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541802 | CR519995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541802 | NM_001112715.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136855 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121726 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158557 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06320674 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06042795 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06042798 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06042802 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06042799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25586013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25586000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25586005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25586011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25586010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25586003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25586007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06042798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06246094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06246086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06088165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06361346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06204829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06088160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06042796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06556677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06388205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06088166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06556673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585979 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558771 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558770 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558772 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570435 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424254_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558769 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570434 | - | - |
| Mouse Genome 430 2.0 Array | 1424254_at | - | - |
| Mouse Genome 430 2.0 Array | 1424254_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68713_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68713_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700017910.mm.1 | TC0700002034.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017910.mm.1 | NM_001112715.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017910.mm.1 | NM_001112715.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017911.mm.1 | TC0700002034.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017911.mm.1 | NM_001112715.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017911.mm.1 | NM_001112715.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017914.mm.1 | TC0700002034.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017914.mm.1 | NM_001112715.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017914.mm.1 | NM_001112715.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017914.mm.1 | NM_026820.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017914.mm.1 | NM_026820.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017906.mm.1 | TC0700002034.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017906.mm.1 | NM_026820.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017906.mm.1 | NM_026820.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017913.mm.1 | TC0700002034.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017913.mm.1 | NM_001112715.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017913.mm.1 | NM_001112715.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017913.mm.1 | NM_026820.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017913.mm.1 | NM_026820.3 | Refseq |
| Mouse Expression Set 430 | 1424254_at | - | - |
| Mouse Expression Set 430 | 1424254_PM_at | - | - |
| Murine Genome U74v2 Set | 160254_PM_at | - | - |
| Murine Genome U74v2 Set | 160254_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424254_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424254_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424254_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424254_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 307562 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6510075 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3120438 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3120438 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1690601 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6510075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103140402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001690601 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006510075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003120438 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217180 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2640766 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245129 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2640766 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217180 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214071 | - | - |
| mouse-6 v1.1 (Illumina) | GI_40254488-S | - | - |
| mouse-6 v1.1 (Illumina) | scl068713.2_9-S | - | - |
| mouse-6 v1.1 (Illumina) | scl31899.2_128-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0068713.1_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3120438 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103140402 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6510075 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1690601 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31899.2_128-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068713.1_6-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068713.2_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3120438 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103140402 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6510075 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2640765 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2814484 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2640765 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245129 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217180 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214071 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2814484 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068713.2_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31899.2_128-S | - | - |

Ifitm1
Genomic Map