Assay Details
| Gene Symbol: Serpina3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011458.2 | NP_035588.2 | 3 - 4 | 992 | 67 | |
| GenBank mRNA | AK004842.1 | - | 3 - 4 | 989 | 67 | |
| AK149436.1 | - | 3 - 4 | 993 | 67 | ||
| X56786.1 | - | 3 - 4 | 981 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P319093 | X56786.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P319093 | NM_011458.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P319093 | AK004842.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P319093 | AK149436.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P319093 | X56786.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P319093 | NM_011458.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P319093 | AK004842.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P319093 | AK149436.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456988 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107357 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P319093 | X56786.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P319093 | NM_011458.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P319093 | AK004842.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P319093 | AK149436.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P319093 | X56786.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P319093 | NM_011458.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P319093 | AK004842.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P319093 | AK149436.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702720 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02595125 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02595116 | - | - |
| Whole Mouse Genome G4122A | A_52_P319093 | X56786.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319093 | NM_011458.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319093 | AK004842.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319093 | AK149436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319093 | X56786.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319093 | NM_011458.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319093 | AK004842.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319093 | AK149436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240036 | D00725.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02595116 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10167755 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10167753 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10167751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10357084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10279108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10167755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10290134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10117753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10117755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10290103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10117750 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608675 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423866_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423867_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398060 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608675 | - | - |
| Mouse Genome 430 2.0 Array | 1423867_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423866_at | - | - |
| Mouse Genome 430 2.0 Array | 1457263_at | - | - |
| Mouse Genome 430 2.0 Array | 1457263_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423867_at | - | - |
| Mouse Genome 430 2.0 Array | 1423866_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20714_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20714_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007473.mm.1 | NM_011458.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007473.mm.1 | NM_011458.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007473.mm.1 | TC1200001044.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423866_at | - | - |
| Mouse Expression Set 430 | 1423867_at | - | - |
| Mouse Expression Set 430 | 1423867_PM_at | - | - |
| Mouse Expression Set 430 | 1423866_PM_at | - | - |
| Murine Genome U74v2 Set | 92583_PM_at | - | - |
| Murine Genome U74v2 Set | 92583_at | - | - |
| Murine Genome U74v2 Set | 168983_PM_at | - | - |
| Murine Genome U74v2 Set | 168983_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423867_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423866_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423867_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423866_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423866_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423866_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423867_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423867_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 470968 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2970494 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2970494 | - | - |

Serpina3k
Genomic Map