Assay Details
| Gene Symbol: Sat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026991.2 | NP_081267.1 | 3 - 4 | 360 | 99 | |
| GenBank mRNA | AK011406.1 | - | 4 - 5 | 279 | 99 | |
| AK161230.1 | - | 3 - 4 | 418 | 99 | ||
| BC061227.1 | - | 3 - 4 | 360 | 30312881 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sat2
Genomic Map