Assay Details
| Gene Symbol: Mir485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030253.1 | 1 | 35 | 61 | ||
| Gene Symbol: Mirg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU434920.1 | - | 1703 | 61 | ||
| EU616812.1 | - | 2540 | 61 | |||
| EU616813.1 | - | 2453 | 61 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 668 | Mir668 |
Gm27556, Mirn668, mir-668, mmu-mir-668 |
-105.0 | Sense | Chr. 12 - 109734732 - 109734797 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir485
Genomic Map