Assay Details
Gene Symbol: Cyp26c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105201.1 | NP_001098671.1 | 5 - 6 | 1175 | 98 | |
GenBank mRNA | BC151098.1 | - | 5 - 6 | 1274 | 9088011 | 98 |
BC151106.1 | - | 5 - 6 | 1274 | 9088019 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035407 | NM_001105201.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035407 | BC151098.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035407 | BC151106.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907220 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250250 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409882 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65951 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035407 | NM_001105201.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035407 | BC151098.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035407 | BC151106.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32096995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32097013 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096967 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14506653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096996 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097012 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097029 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14331165 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14147508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096976 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097003 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14147520 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14147547 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14331181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14331157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14506652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097037 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14147498 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14226126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14147496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097015 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097033 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14147522 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14147519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097017 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097018 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14506651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32096975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097008 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14506661 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32097042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14209603 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10462849 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462847 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462851 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462852 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462850 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462848 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10462846 | - | - |
Affymetrix Mouse Clariom S Array | PSR1900004825.mm.1 | TC1900000537.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900004824.mm.1 | NM_001105201.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900004824.mm.1 | NM_001105201.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900004824.mm.1 | TC1900000537.mm.1 | MTA TC |
