Assay Details
| Gene Symbol: Ak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027592.3 | NP_081868.1 | 2 - 3 | 196 | 99 | |
| GenBank mRNA | AK014903.1 | - | 1 - 2 | 118 | 99 | |
| BC019453.1 | - | 2 - 3 | 147 | 4024616 | 99 | |
More Information
Set Membership: |
|

Ak6
Genomic Map