Assay Details
| Gene Symbol: Ak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027592.3 | NP_081868.1 | 4 - 5 | 396 | 152 | |
| GenBank mRNA | AK012905.1 | - | 4 - 5 | 346 | 152 | |
| AK014903.1 | - | 3 - 4 | 318 | 152 | ||
| BC019453.1 | - | 4 - 5 | 347 | 4024616 | 152 | |
| BG084768.2 | - | 2 - 3 | 183 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ak6
Genomic Map