Assay Details
| Gene Symbol: Ftx | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028381.1 | 3 - 4 | 505 | 82 | |||
| GenBank mRNA | AK020989.1 | - | LIT1001 Ftx |
3 - 4 | 372 | 82 | |
| AK035525.1 | - | 3 - 4 | 507 | 82 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 374b | Mir374b |
Mir374, Mirn374, mir-374b, mmu-mir-374, mmu-mir-374b |
-36.2kb | Sense | Chr. X - 103573060 - 103573154 on NCBI Build GRCm38 |
| Downstream |
zinc finger, CCHC domain containing 13 |
Zcchc13 |
4930513O09Rik, Cnbp2 |
17.4kb | Anti-sense | Chr. X - 103630586 - 103631664 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Ftx
Genomic Map