Assay Details
| Gene Symbol: Ftx | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028380.1 | 4 - 5 | 600 | 108 | |||
| NR_028381.1 | 4 - 5 | 621 | 108 | ||||
| GenBank mRNA | AK020989.1 | - | LIT1001 Ftx |
4 - 5 | 488 | 108 | |
| AK035525.1 | - | 4 - 5 | 623 | 108 | |||
| AK085125.1 | - | 4 - 5 | 487 | 109 | |||
| BC027389.1 | - | 3 - 4 | 396 | 108 | |||
| BC076616.1 | - | 4 - 5 | 509 | 108 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 374b | Mir374b |
Mir374, Mirn374, mir-374b, mmu-mir-374, mmu-mir-374b |
-34.5kb | Sense | Chr. X - 103573060 - 103573154 on NCBI Build GRCm38 |
| Downstream |
zinc finger, CCHC domain containing 13 |
Zcchc13 |
4930513O09Rik, Cnbp2 |
21.2kb | Anti-sense | Chr. X - 103630586 - 103631664 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175002 | AK085125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175002 | BC027389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175002 | BC076616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175002 | NR_028380.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175002 | NR_028381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175007 | AK085125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175007 | BC027389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175007 | BC076616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175007 | NR_028380.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175007 | NR_028381.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399966 | AK081630.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 321307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915198 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918884 | AB001750.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918884 | AK134587.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918884 | AK163126.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918884 | BC003921.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918884 | BC060166.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918884 | BC080274.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918884 | D86066.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918884 | NM_019400.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837337 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524098 | AK043578.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524098 | AK043659.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257006 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441895 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129972 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125203 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165095 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111409 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175436 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175002 | AK085125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175002 | BC027389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175002 | BC076616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175002 | NR_028380.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175002 | NR_028381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175007 | AK085125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175007 | BC027389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175007 | BC076616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175007 | NR_028380.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175007 | NR_028381.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09380541 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03729735 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09380492 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03729832 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03729758 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09380572 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175436 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09380527 | - | - |
| Whole Mouse Genome G4122A | A_52_P177865 | NR_028381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177865 | AK035525.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128861 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09380572 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175436 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03729758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15181109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14878466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14996939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14764507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14712238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14747659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14922703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14996938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15003401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14747657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14817889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15126523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15126515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32573780 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1438838_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438838_at | - | - |
| Mouse Genome 430 2.0 Array | 1438838_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00017553.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017553.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017553.mm.1 | NR_028381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017553.mm.1 | NR_028381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017553.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017552.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017552.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017552.mm.1 | NR_028381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017552.mm.1 | NR_028381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017552.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017540.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017540.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017540.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017539.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017539.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017539.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017548.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017548.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017548.mm.1 | NR_028381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017548.mm.1 | NR_028381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017548.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017537.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017537.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017537.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017535.mm.1 | NR_028381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017535.mm.1 | NR_028381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017535.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017541.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017541.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017541.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017534.mm.1 | NR_028381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017534.mm.1 | NR_028381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017534.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017546.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017546.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017546.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017547.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017547.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017547.mm.1 | NR_028381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017547.mm.1 | NR_028381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017547.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017550.mm.1 | NR_028380.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017550.mm.1 | NR_028380.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017550.mm.1 | NR_028381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017550.mm.1 | NR_028381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017550.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00017536.mm.1 | NR_028381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017536.mm.1 | NR_028381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017536.mm.1 | TC0X00002778.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438838_at | - | - |
| Mouse Expression Set 430 | 1438838_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438838_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438838_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 688037 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102120279 | - | - |

Ftx
Genomic Map