Assay Details
| Gene Symbol: D2Ertd173e | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083486.1 | - | LIT1672 Nespas |
1 | 3653 | 138 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 298 | Mir298 |
Mirn298, mir-298, mmu-mir-298 |
-9.0kb | Sense | Chr. 2 - 174267504 - 174267585 on NCBI Build GRCm38 |
| Downstream | neuroendocrine secretory protein antisense | Nespas |
Gnas-as, Nespos |
877.0 | Sense | Chr. 2 - 174281237 - 174295436 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D2Ertd173e
Genomic Map