Assay Details
| Gene Symbol: Kis2 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY940615.1 | - | LIT3388 Kis2 |
1 - 2 | 358 | 59 | |
| AY940618.1 | - | LIT3391 Kis2 |
1 - 2 | 358 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 18b | Mir18b |
Mirn18b, mir-18b, mmu-mir-18b |
-150.0 | Sense | Chr. X - 52742331 - 52742413 on NCBI Build GRCm38 |
| Downstream | coiled-coil domain containing 160 | Ccdc160 | 6330534C20Rik | 46.6kb | Anti-sense | Chr. X - 52791200 - 52799474 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Kis2
Genomic Map