Assay Details
| Gene Symbol: Pinc | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003202.1 | 3 - 4 | 350 | 82 | |||
| GenBank mRNA | DQ059755.1 | - | LIT3460 PINC |
3 - 4 | 350 | 82 | |
| DQ059756.1 | - | LIT3461 PINC |
4 - 5 | 514 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D530049I02 gene | D530049I02Rik | Anti-sense | Chr. 1 - 73398968 - 73430779 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA 6030407O03 gene | 6030407O03Rik | 218.1kb | Sense | Chr. 1 - 73618381 - 73864466 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Pinc
Genomic Map