Assay Details
| Gene Symbol: Pinc | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003202.1 | 4 - 5 | 542 | 69 | |||
| GenBank mRNA | DQ059755.1 | - | LIT3460 PINC |
4 - 5 | 542 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700027A15 gene | 1700027A15Rik | -365.9kb | Anti-sense | Chr. 1 - 73016036 - 73025508 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA D530049I02 gene | D530049I02Rik | 1.8kb | Anti-sense | Chr. 1 - 73398968 - 73430779 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179186 | DQ059755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179186 | NR_003202.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735126 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 6468 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179186 | DQ059755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179186 | NR_003202.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04145265 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00102361 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00102361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00828064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00225903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00667079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00225889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00136784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01115122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00136782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00840301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00920010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00034425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00053502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01090344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00034422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00225870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00628101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00174304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00390396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00628103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00174301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00225907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00463353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00808788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00667083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00920006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20342519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00920005 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0100020682.mm.1 | NR_003202.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100020682.mm.1 | NR_003202.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100020682.mm.1 | TC0100002569.mm.1 | MTA TC |

Pinc
Genomic Map