Assay Details
| Gene Symbol: Ipw | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015351.1 | 315 | 86 | ||||
| GenBank mRNA | AK045353.1 | - | 3 - 4 | 2279 | 86 | ||
| U69888.1 | - | LIT1675 IPW |
315 | 86 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA C230091D08 gene | C230091D08Rik | -345.4kb | Anti-sense | Chr. 7 - 59307924 - 59324149 on NCBI Build GRCm38 | |
| Downstream |
small nucleolar RNA, C/D box 116 |
Snord116 |
MBII-85, Pwcr1 |
2.1kb | Anti-sense | Chr. 7 - 59675990 - 59676079 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Ipw
Genomic Map