Assay Details
| Gene Symbol: Meg3 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027651.2 | 3 - 4 | 925 | 69 | |||
| NR_027652.1 | 3 - 4 | 969 | 69 | ||||
| GenBank mRNA | BC024818.1 | - | 1 - 2 | 297 | 69 | ||
| BC094421.1 | - | 3 - 4 | 914 | 69 | |||
| Y13832.1 | - | LIT3453 Gtl2 |
3 - 4 | 960 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1906-1 | Mir1906-1 |
Mir1906, Mirn1906, mmu-mir-1906, mmu-mir-1906-1 |
Sense | Chr. 12 - 109544541 - 109544620 on NCBI Build GRCm38 | |
| Downstream | microRNA 770 | Mir770 |
Mirn770, mir-770, mmu-mir-770 |
18.7kb | Sense | Chr. 12 - 109563692 - 109563785 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Meg3
Genomic Map