Assay Details
| Gene Symbol: Rn7sk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030687.1 | 1 | 249 | 143 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | intestinal cell kinase | Ick |
2210420N10Rik, AI848300, Mrk |
-3.2kb | Anti-sense | Chr. 9 - 78109192 - 78172114 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA C920006O11 gene | C920006O11Rik | 281.0 | Anti-sense | Chr. 9 - 78175914 - 78178882 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rn7sk
Genomic Map