Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26644 |
Gm26644 | -79.9kb | Sense | Chr. 19 - 44685753 - 44695379 on NCBI Build GRCm38 | |
| Downstream | microRNA 6405 | Mir6405 |
Gm27349, mmu-mir-6405 |
23.9kb | Anti-sense | Chr. 19 - 44808406 - 44808529 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map