Assay Details
| Gene Symbol: Fgfr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163215.2 | NP_001156687.1 | 7 | 1112 | 87 | |
| NM_001163216.2 | NP_001156688.1 | 5 | 827 | 87 | ||
| NM_001163217.2 | NP_001156689.1 | 6 | 881 | 87 | ||
| NM_001205270.1 | NP_001192199.1 | 7 | 1086 | 87 | ||
| NM_008010.5 | NP_032036.2 | 7 | 1086 | 87 | ||
| GenBank mRNA | AF024638.1 | - | 5 | 735 | 87 | |
| AK140217.1 | - | 6 | 1003 | 87 | ||
| AK163495.1 | - | 5 | 665 | 87 | ||
| AK163547.1 | - | 6 | 1003 | 87 | ||
| BC053056.1 | - | 7 | 996 | 5708838 | 87 | |
| BU053488.1 | - | 2 | 102 | 87 | ||
| M81342.1 | - | 7 | 1004 | 87 | ||
| S56291.1 | - | 6 | 831 | 87 | ||
| X58255.1 | - | 7 | 990 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fgfr3
Genomic Map