Assay Details
| Gene Symbol: Gm12942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099319.2 | NP_001092789.1 | 1 - 2 | 148 | 86 | |
| GenBank mRNA | AK156456.1 | - | 1 - 2 | 130 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gm12942
Genomic Map