Assay Details
| Gene Symbol: Sec14l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127725.1 | NP_001121197.1 | 7 - 8 | 843 | 94 | |
| GenBank mRNA | BC157907.1 | - | 7 - 8 | 909 | 9088109 | 94 |
| BC157990.1 | - | 7 - 8 | 843 | 9007304 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l5
Genomic Map