Assay Details
| Gene Symbol: Plcxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134480.1 | NP_001127952.1 | 3 - 4 | 1428 | 115 | |
| GenBank mRNA | BC151059.1 | - | 3 - 4 | 1030 | 9087972 | 115 |
| BC151063.1 | - | 3 - 4 | 1030 | 9087976 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045755 | BC064457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045755 | NM_001134480.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045755 | BC049364.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695932 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625566 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 81165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 698717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820114 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84016 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157886 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045755 | BC064457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045755 | NM_001134480.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045755 | BC049364.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476492 | BC064457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476492 | NM_001134480.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476492 | AK086534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476492 | BC049364.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P628997 | BC151063.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P628997 | BC151059.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P628997 | NM_001134480.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12619034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12831325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12754622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12831313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12754629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12754606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12773882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12831317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12926171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12754620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30729641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12562240 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10439739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439738 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455324_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435462_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10439732 | - | - |
| Mouse Genome 430 2.0 Array | 1455324_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435462_at | - | - |
| Mouse Genome 430 2.0 Array | 1435462_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455324_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 433022_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 433022_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600013704.mm.1 | NM_001134480.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013704.mm.1 | NM_001134480.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013704.mm.1 | TC1600001671.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600013705.mm.1 | NM_001134480.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013705.mm.1 | NM_001134480.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013705.mm.1 | TC1600001671.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600013703.mm.1 | NM_001134480.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013703.mm.1 | NM_001134480.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013703.mm.1 | TC1600001671.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600013706.mm.1 | NM_001134480.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013706.mm.1 | NM_001134480.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013706.mm.1 | TC1600001671.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455324_at | - | - |
| Mouse Expression Set 430 | 1435462_at | - | - |
| Mouse Expression Set 430 | 1455324_PM_at | - | - |
| Mouse Expression Set 430 | 1435462_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435462_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435462_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455324_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455324_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 468885 | - | - |

Plcxd2
Genomic Map