Assay Details
| Gene Symbol: Aph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146104.3 | NP_666216.1 | 5 - 6 | 861 | 95 | |
| NM_146134.2 | NP_666246.1 | 5 - 6 | 861 | 95 | ||
| GenBank mRNA | AK050828.1 | - | 3 - 4 | 1084 | 95 | |
| AK078343.1 | - | 3 - 4 | 326 | 95 | ||
| AK148400.1 | - | 5 - 6 | 814 | 95 | ||
| AK152334.1 | - | 5 - 6 | 804 | 95 | ||
| AK178736.1 | - | 2 - 3 | 155 | 96 | ||
| AK182589.1 | - | 2 - 3 | 155 | 95 | ||
| AK190873.1 | - | 2 - 3 | 155 | 95 | ||
| AK216088.1 | - | 2 - 3 | 155 | 95 | ||
| BC012406.1 | - | 5 - 6 | 786 | 3483214 | 95 | |
| BC024111.1 | - | 5 - 6 | 727 | 5098973 | 95 | |
| BC057865.1 | - | 5 - 6 | 804 | 5320175 | 95 | |
| CX207467.1 | - | 1 - 2 | 48 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aph1a
Genomic Map