Assay Details
| Gene Symbol: Pcdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198117.2 | NP_932785.1 | 1 - 2 | 2502 | 65 | |
| GenBank mRNA | AY013760.1 | - | 1 - 2 | 2389 | 65 | |
| BC030897.1 | - | 1 - 2 | 2502 | 65 | ||
| BC141390.1 | - | 1 - 2 | 2483 | 9056003 | 65 | |
| EU854475.1 | - | 1 - 2 | 1492 | 65 | ||
More Information
Set Membership: |
|

Pcdha2
Genomic Map