Assay Details
| Gene Symbol: Speer4cos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001585.3 | 1 - 2 | 135 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | spermatogenesis associated glutamate (E)-rich protein pseudogene | 4930572O03Rik | -23.7kb | Anti-sense | Chr. 5 - 15652288 - 15657059 on NCBI Build GRCm38 | |
| Downstream | spermatogenesis associated glutamate (E)-rich protein 4c | Speer4c |
4933431D05Rik, SPEER-4C |
10.3kb | Anti-sense | Chr. 5 - 15709505 - 15714675 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Speer4cos
Genomic Map