Assay Details
| Gene Symbol: Snord15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002172.1 | 1 | 74 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S3 | Rps3 |
D7Ertd795e, Rs_3 |
Sense | Chr. 7 - 99477896 - 99483709 on NCBI Build GRCm38 | |
| Downstream |
arrestin, beta 1 |
Arrb1 |
1200006I17Rik, AW208571, G430100A01Rik |
52.6kb | Anti-sense | Chr. 7 - 99535486 - 99606772 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord15a
Genomic Map