Assay Details
| Gene Symbol: BC005561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166581.1 | NP_001160053.1 | 2 | 5080 | 132 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polycystic kidney disease 2 | Pkd2 |
C030034P18Rik, PC2, TRPP2 |
-11.6kb | Sense | Chr. 5 - 104457224 - 104505819 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA D930016D06 gene | D930016D06Rik |
100041172, 1110004B06Rik, AI451450, Gm3179 |
3.4kb | Sense | Chr. 5 - 104525735 - 104554211 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BC005561
Genomic Map