Assay Details
| Gene Symbol: Gm6548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003363.1 | 1 | 249 | 108 | ||
| GenBank mRNA | BC089461.1 | - | 1 | 249 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G patch domain containing 11 | Gpatch11 |
2310002B06Rik, C80922, Ccdc75, L26697 |
-2.2kb | Anti-sense | Chr. 17 - 78835516 - 78848308 on NCBI Build GRCm38 |
| Downstream | eukaryotic translation initiation factor 2-alpha kinase 2 | Eif2ak2 |
2310047A08Rik, 4732414G15Rik, AI467567, AI747578, Pkr, Prkr, Tik |
6.0 | Sense | Chr. 17 - 78852550 - 78882572 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm6548
Genomic Map