Assay Details
| Gene Symbol: H2afy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003523.1 | 1 | 52 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | plasmacytoma variant translocation 1 | Pvt1 |
Gt(pU21)84Imeg, Mis-1, Mlvi-1, Pvt-1 |
Anti-sense | Chr. 15 - 62037987 - 62250975 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 30159 |
Gm30159 | 891.0kb | Sense | Chr. 15 - 63110465 - 63145446 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

H2afy3
Genomic Map