Assay Details
| Gene Symbol: Vax2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002871.1 | 1 - 2 | 99 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CD207 antigen | Cd207 | -32.9kb | Sense | Chr. 6 - 83671207 - 83677857 on NCBI Build GRCm38 | |
| Downstream |
ATPase, H+ transporting, lysosomal V1 subunit B1 |
Atp6v1b1 |
AW208839, Atp6b1, D630003L15, D630030L16Rik, D630039P21Rik, Vpp-3, Vpp3 |
30.8kb | Anti-sense | Chr. 6 - 83743017 - 83758810 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Vax2os
Genomic Map