Assay Details
| Gene Symbol: Vax2os | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002873.2 | 1 - 2 | 195 | 100 | |||
| NR_075094.1 | 1 - 2 | 69 | 100 | ||||
| NR_075095.1 | 1 - 2 | 195 | 100 | ||||
| GenBank mRNA | AK044313.1 | - | LIT3405 Vax2OS |
1 - 2 | 70 | 100 | |
| AY589787.1 | - | LIT3407 Vax2OS |
1 - 2 | 195 | 100 | ||
| AY589788.1 | - | LIT3408 Vax2OS |
1 - 2 | 195 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CD207 antigen | Cd207 | -24.2kb | Sense | Chr. 6 - 83671207 - 83677857 on NCBI Build GRCm38 | |
| Downstream | ventral anterior homeobox 2 | Vax2 | Dres93 | 332.0 | Anti-sense | Chr. 6 - 83711264 - 83739229 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Vax2os
Genomic Map