Assay Details
| Gene Symbol: Kcnq1ot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001461.5 | 1 | 81151 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transient receptor potential cation channel, subfamily M, member 5 |
Trpm5 |
9430099A16Rik, LTrpC-5, Ltrpc5, Mtr1 |
-118.5kb | Sense | Chr. 7 - 143071529 - 143094642 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933417O13 gene | 4933417O13Rik | 26.2kb | Sense | Chr. 7 - 143322772 - 143330650 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kcnq1ot1
Genomic Map